cis-4-Methylcyclohexanol ≥98.0%

Supplier: TCI America

M0353-5ML M0353-1ML
TCM0353-005MLEA 189.6 CAD
TCM0353-005ML TCM0353-1ML
cis-4-Methylcyclohexanol ≥98.0%
cis-4-Methylcyclohexanol

CAS Number: 7731-28-4
MDL Number: MFCD00064170
Molecular Formula: C7H14O
Molecular Weight: 114.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 171
Flash Point (°C): 70
Specific Gravity (20/20): 0.93

Formula: C₇H₁₄O
Boiling Pt: 171 °C
Melting Pt: –41 °C
Density: 0.93 (20 °C)
Flash Pt: 70 °C
MDL Number: MFCD00064170
CAS Number: 7731-28-4
EINECS: 231-789-1

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR