5-Methylindole ≥99.0%

Supplier: TCI America
Warning

M0348-1G M0348-5G
TCM0348-001GEA 94.8 CAD
TCM0348-001G TCM0348-5G
5-Methylindole ≥99.0%
5-Methylindole

CAS Number: 614-96-0
MDL Number: MFCD00005680
Molecular Formula: C9H9N
Molecular Weight: 131.18
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 100
Melting point (°C): 60
Storage Temperature: 0-10°C

Formula: C₉H₉N
Boiling Pt: 100 °C
Melting Pt: 60 °C
Density: 1.0202 g/cm³ (78.3 °C)
MDL Number: MFCD00005680
CAS Number: 614-96-0
EINECS: 210-400-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=766616918ca8191c9d7b6c9e5f2e7907961a4cc1&vid=766616918ca8191c9d7b6c9e5f2e7907961a4cc1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR