3-Methylindole ≥98.0%

Supplier: TCI America

M0347-5G M0347-25G M0347-1G
TCM0347-5GEA 62.88 CAD
TCM0347-5G TCM0347-025G TCM0347-001G
3-Methylindole ≥98.0%
3-Methylindole

CAS Number: 83-34-1
MDL Number: MFCD00005627
Molecular Formula: C9H9N
Molecular Weight: 131.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 141
Melting point (°C): 96
Flash Point (°C): 132


Other Product Names / Synonyms: Skatole

Formula: C₉H₉N
Boiling Pt: 141 °C
Melting Pt: 96 °C
Density: 1.003 g/cm³ (99.6 °C)
Flash Pt: 132 °C
MDL Number: MFCD00005627
CAS Number: 83-34-1
EINECS: 201-471-7
Merck Index: 12,08700
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75dbbf9f8a80be304b13d241c05541519c183458&vid=75dbbf9f8a80be304b13d241c05541519c183458&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR