2-Methylindole ≥99.0%
Supplier: TCI America
|
Warning
|
M0346-500G
M0346-25G
TCM0346-500GEA
816.86
CAD
TCM0346-500G
TCM0346-025G
2-Methylindole ≥99.0%
2-Methylindole
CAS Number: 95-20-5
MDL Number: MFCD00005616
Molecular Formula: C9H9N
Molecular Weight: 131.18
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 272
Melting point (°C): 60
Flash Point (°C): 141
Product Categories: Synthetic Organic Chemistry, Peptide Synthesis
Other Product Names / Synonyms: Methylketole
Formula:
C₉H₉N Boiling Pt: 272 °C Melting Pt: 60 °C Density: 1.07 g/cm³ (20 °C) Flash Pt: 141 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005616 CAS Number: 95-20-5 EINECS: 202-398-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...