2-Methylhydroquinone ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Methylhydroquinone

M0342-25G M0342-500G M0342-100G
TCM0342-025GEA 45.82 CAD
TCM0342-025G TCM0342-500G TCM0342-100G
2-Methylhydroquinone ≥98.0% (by titrimetric analysis)
2-Methylhydroquinone

CAS Number: 95-71-6
MDL Number: MFCD00002345
Molecular Formula: C7H8O2
Molecular Weight: 124.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 126
Flash Point (°C): 172


Other Product Names / Synonyms: 2,5-Dihydroxytoluene, p-Toluhydroquinone

Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 126 °C
Flash Pt: 172 °C
Storage Temperature: Ambient
MDL Number: MFCD00002345
CAS Number: 95-71-6
EINECS: 202-443-7
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f94420ed7877288a6dcbfec8ff27d4bc946693ea&vid=f94420ed7877288a6dcbfec8ff27d4bc946693ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR