Sarcosine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

M0332-500G M0332-25G M0332-100G
TCM0332-500GEA 284.24 CAD
TCM0332-500G TCM0332-025G TCM0332-100G
Sarcosine ≥98.0% (by titrimetric analysis)
Sarcosine

CAS Number: 107-97-1
MDL Number: MFCD00004279
Molecular Formula: C3H7NO2
Molecular Weight: 89.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 210


Product Categories: Bioscience, Biologically-modified Amino Acids
Other Product Names / Synonyms: Methylaminoacetic Acid, N-Methylglycocoll, N-Methylglycine, Me-Gly-OH

Formula: C₃H₇NO₂
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 210 °C
Density: 1.301 g/cm³ (20 °C)
MDL Number: MFCD00004279
CAS Number: 107-97-1
EINECS: 203-538-6
Merck Index: 12,08519
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29ebb0d7572f52a06b622d3839c45ceb21a210fb&vid=29ebb0d7572f52a06b622d3839c45ceb21a210fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR