5-Methyl-1,10-phenanthroline
Supplier: TCI America
M0300-1G
M0300-100MG
TCM0300-001GEA
363.72
CAD
TCM0300-001G
TCM0300-100MG
5-Methyl-1,10-phenanthroline
5-Methyl-1,10-phenanthroline
[for Colorimetric Determination of Iron]
CAS Number: 3002-78-6
MDL Number: MFCD00004982
Molecular Formula: C13H10N2
Molecular Weight: 194.24
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 113
Formula:
C₁₃H₁₀N₂ Boiling Pt: 351 °C (1013 hPa) Melting Pt: 113 °C |
MDL Number:
MFCD00004982 CAS Number: 3002-78-6 EINECS: 221-099-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...