3-Methylbiphenyl ≥95.0%
Supplier: TCI America
M0296-1G
M0296-5G
M0296-25G
TCM0296-001GEA
90.06
CAD
TCM0296-001G
TCM0296-005G
TCM0296-25G
3-Methylbiphenyl ≥95.0%
3-Methylbiphenyl
CAS Number: 643-93-6
MDL Number: MFCD00008533
Molecular Formula: C13H12
Molecular Weight: 168.24
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 272
Flash Point (°C): 110
Specific Gravity (20/20): 1.01
Other Product Names / Synonyms: 3-Phenyltoluene
Formula:
C₁₃H₁₂ Boiling Pt: 272 °C Melting Pt: 4.53 °C Density: 1.01 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00008533 CAS Number: 643-93-6 EINECS: 211-404-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...