2-Methylbenzimidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Methyl-1H-benzo[d]imidazole, 2-Methyl-1H-benzimidazole

M0286-25G M0286-250G
TCM0286-025GEA 42.66 CAD
TCM0286-025G TCM0286-250G
2-Methylbenzimidazole ≥98.0% (by GC, titration analysis)
2-Methylbenzimidazole

CAS Number: 615-15-6
MDL Number: MFCD00005598
Molecular Formula: C8H8N2
Molecular Weight: 132.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 175

Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 339
Melting Pt: 175
MDL Number: MFCD00005598
CAS Number: 615-15-6
EINECS: 210-411-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc3985c69babef7be38ad51f6e636025dae6b995&vid=bc3985c69babef7be38ad51f6e636025dae6b995&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR