4-(Methylamino)benzoic acid ≥98.0%
Supplier: TCI America
M0267-5G
M0267-25G
TCM0267-005GEA
153.26
CAD
TCM0267-005G
TCM0267-025G
4-(Methylamino)benzoic acid ≥98.0%
4-(Methylamino)benzoic acid
CAS Number: 10541-83-0
MDL Number: MFCD00002535
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 161
Formula:
C₈H₉NO₂ MW: 151.16 g/mol Boiling Pt: 296 Melting Pt: 159 |
MDL Number:
MFCD00002535 CAS Number: 10541-83-0 EINECS: 234-121-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...