5-Methyl-2-furaldehyde ≥98.0%

Supplier: TCI America

M0254-25ML M0254-500ML
TCM0254-025MLEA 50.56 CAD
TCM0254-025ML TCM0254-500ML
5-Methyl-2-furaldehyde ≥98.0%
5-Methyl-2-furaldehyde

CAS Number: 620-02-0
MDL Number: MFCD00003232
Molecular Formula: C6H6O2
Molecular Weight: 110.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 187
Flash Point (°C): 72
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: 5-Methyl-2-furfural

Formula: C₆H₆O₂
Boiling Pt: 187 °C
Density: 1.11 (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00003232
CAS Number: 620-02-0
EINECS: 210-622-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e508c86b3994b5a4e84399a78331e45165d9b55&vid=9e508c86b3994b5a4e84399a78331e45165d9b55&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR