(Methyl)triphenylphosphonium iodide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Methyltriphenylphosphonium iodide

M0253-25G M0253-500G M0253-100G
TCM0253-025GEA 89.43 CAD
TCM0253-025G TCM0253-500G TCM0253-100G
(Methyl)triphenylphosphonium iodide ≥98.0% (by titrimetric analysis)
(Methyl)triphenylphosphonium iodide

CAS Number: 2065-66-9
MDL Number: MFCD00066175
Molecular Formula: C19H18IP
Molecular Weight: 404.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 185

Formula: C₁₉H₁₈IP
MW: 404.23 g/mol
Melting Pt: 185 °C
MDL Number: MFCD00066175
CAS Number: 2065-66-9
EINECS: 218-178-5
UN: 3464
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=914fbd0e26a606e5fc9ff3d4f98c837b2b37577e&vid=914fbd0e26a606e5fc9ff3d4f98c837b2b37577e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR