2-Pyridinethiol ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
M0246-25G
M0246-100G
TCM0246-025GEA
117.35
CAD
TCM0246-025G
TCM0246-100G
2-Pyridinethiol ≥98.0% (by HPLC, titration analysis)
2-Pyridinethiol
CAS Number: 2637-34-5
MDL Number: MFCD00006285
Molecular Formula: C5H5NS
Molecular Weight: 111.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 127
Product Categories: Synthetic Organic Chemistry, Thiols
Other Product Names / Synonyms: 2-Pyridinethiol, 2-Pyridyl Mercaptan
Formula:
C₅H₅NS Boiling Pt: 259 °C (1013 hPa) Melting Pt: 127 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006285 CAS Number: 2637-34-5 EINECS: 220-131-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...