D-(-)-N-Methylglucamine ≥99.0% (by titrimetric analysis)

Supplier: TCI America

M0227-25G M0227-500G
TCM0227-025GEA 33.18 CAD
TCM0227-025G TCM0227-500G
D-(-)-N-Methylglucamine ≥99.0% (by titrimetric analysis)
D(-)-N-Methylglucamine

CAS Number: 6284-40-8
MDL Number: MFCD00004707
Molecular Formula: C7H17NO5
Molecular Weight: 195.22
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 131
Specific rotation [a]20/D: -16.5 deg (C=2, H2O)


Product Categories: Bioscience, Sugars (Others), Aminosugars
Other Product Names / Synonyms: 1-Deoxy-1-(methylamino)-D-glucitol

Formula: H₃CNHCH₂(CH(OH))₄CH₂OH
Melting Pt: 131 °C
Storage Temperature: Ambient
MDL Number: MFCD00004707
CAS Number: 6284-40-8
EINECS: 228-506-9
Merck Index: 12,06154
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e12e2a9157cfdf786a4a7ed13fe8357e4c7a840&vid=2e12e2a9157cfdf786a4a7ed13fe8357e4c7a840&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR