Itaconic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: 2-Methylenesuccinic acid, Methylenesuccinic acid

M0223-500G M0223-25G
TCM0223-500GEA 35.34 CAD
TCM0223-500G TCM0223-025G
Itaconic acid ≥99.0% (by titrimetric analysis)
Itaconic acid

CAS Number: 97-65-4
MDL Number: MFCD00004260
Molecular Formula: C5H6O4
Molecular Weight: 130.10
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 167


Other Product Names / Synonyms: Methylenesuccinic Acid, 2-Propene-1,2-dicarboxylic Acid

Formula: C₅H₆O₄
MW: 130.1 g/mol
Boiling Pt: 381 °C (1013 hPa)
Melting Pt: 167 °C
Density: 1.573 g/cm³ (20 °C)
MDL Number: MFCD00004260
CAS Number: 97-65-4
EINECS: 202-599-6
Merck Index: 13,05264
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=986274e9bae058818059db72907a2206e7162ac9&vid=986274e9bae058818059db72907a2206e7162ac9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR