Methyldiphenylamine ≥98.0%
Supplier: TCI America
M0211-25ML
M0211-5ML
TCM0211-025MLEA
218.04
CAD
TCM0211-025ML
TCM0211-5ML
Methyldiphenylamine ≥98.0%
Methyldiphenylamine
CAS Number: 552-82-9
MDL Number: MFCD00041900
Molecular Formula: C13H13N
Molecular Weight: 183.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 295
Flash Point (°C): 110
Specific Gravity (20/20): 1.05
Formula:
C₁₃H₁₃N Boiling Pt: 295 °C Density: 1.05 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00041900 CAS Number: 552-82-9 EINECS: 209-023-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...