3'-Methylacetophenone ≥97.0%

Supplier: TCI America
Warning

M0191-25ML M0191-100ML
TCM0191-025MLEA 133.49 CAD
TCM0191-025ML TCM0191-100ML
3'-Methylacetophenone ≥97.0%
3'-Methylacetophenone

CAS Number: 585-74-0
MDL Number: MFCD00008742
Molecular Formula: C9H10O
Molecular Weight: 134.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 219
Flash Point (°C): 85
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: 3-Acetyltoluene

Formula: C₉H₁₀O
Boiling Pt: 219 °C
Density: 1.01 (20 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00008742
CAS Number: 585-74-0
EINECS: 209-561-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a28a876814720aa09991a0c09e575b60f9119025&vid=a28a876814720aa09991a0c09e575b60f9119025&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR