2'-Methylacetophenone ≥98.0%

Supplier: TCI America

Synonyms: o-Methylacetophenone, Methyl o-tolyl ketone, 1-(o-Tolyl)ethanone

M0172-25ML M0172-500ML
TCM0172-025MLEA 89.27 CAD
TCM0172-025ML TCM0172-500ML
2'-Methylacetophenone ≥98.0%
2'-Methylacetophenone

CAS Number: 577-16-2
MDL Number: MFCD00008734
Molecular Formula: C9H10O
Molecular Weight: 134.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 216
Flash Point (°C): 76
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: 2-Acetyltoluene

Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 216 °C
Density: 1.02 (20 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00008734
CAS Number: 577-16-2
EINECS: 209-408-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=426cb921b05ebd3312192c01be4097714ba59c3f&vid=426cb921b05ebd3312192c01be4097714ba59c3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR