4-Methylbenzyl alcohol ≥99.0%

Supplier: TCI America

M0162-500G M0162-25G M0162-100G
TCM0162-500GEA 628.84 CAD
TCM0162-500G TCM0162-025G TCM0162-100G
4-Methylbenzyl alcohol ≥99.0%
4-Methylbenzyl alcohol

CAS Number: 589-18-4
MDL Number: MFCD00004664
Molecular Formula: C8H10O
Molecular Weight: 122.17
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 116
Melting point (°C): 60


Other Product Names / Synonyms: alpha-Hydroxy-p-xylene

Formula: C₈H₁₀O
Boiling Pt: 116 °C
Melting Pt: 60 °C
Density: 0.978 g/cm³ (72 °C)
MDL Number: MFCD00004664
CAS Number: 589-18-4
EINECS: 209-639-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=304e12348e50363487e9c1131f3ef79930748397&vid=304e12348e50363487e9c1131f3ef79930748397&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR