Methyl-p-benzoquinone ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Methylcyclohexa-2,5-diene-1,4-dione, Methylquinone, Methyl-1,4-benzoquinone

M0159-5G M0159-25G
TCM0159-005GEA 115.75 CAD
TCM0159-005G TCM0159-025G
Methyl-p-benzoquinone ≥99.0% (by GC, titration analysis)
Methyl-p-benzoquinone

CAS Number: 553-97-9
MDL Number: MFCD00001603
Molecular Formula: C7H6O2
Molecular Weight: 122.12
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 69
Flash Point (°C): 81


Other Product Names / Synonyms: Methyl-p-benzoquinone, Methyl-p-quinone

Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 187 °C (1013 hPa)
Melting Pt: 69 °C
Density: 1.08 g/cm³ (75.5 °C)
Flash Pt: 81 °C
MDL Number: MFCD00001603
CAS Number: 553-97-9
EINECS: 209-056-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c456f94d764ce8371e1292152f07c247d3d49bf&vid=8c456f94d764ce8371e1292152f07c247d3d49bf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR