4'-Methylacetophenone ≥95.0%

Supplier: TCI America
Warning

M0135-25ML M0135-500ML
TCM0135-025MLEA 26.86 CAD
TCM0135-025ML TCM0135-500ML
4'-Methylacetophenone ≥95.0%
4'-Methylacetophenone

CAS Number: 122-00-9
MDL Number: MFCD00008751
Molecular Formula: C9H10O
Molecular Weight: 134.18
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 226
Flash Point (°C): 92
Specific Gravity (20/20): 1.01


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Acetyltoluene

Formula: C₉H₁₀O
Boiling Pt: 226 °C
Melting Pt: 28 °C
Density: 1.01 (20 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00008751
CAS Number: 122-00-9
EINECS: 204-514-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1a43acb52af65f8e0c9f2ad48973bcfa2a6bd64&vid=e1a43acb52af65f8e0c9f2ad48973bcfa2a6bd64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR