Hydroquinone monomethyl ether ≥99.0%

Supplier: TCI America
Warning

Synonyms: p-Methoxyphenol, Mequinol, 4-Methoxyphenol, 1-Hydroxy-4-methoxybenzene, 4-Hydroxyanisole

M0123-25G M0123-500G M0123-100G
TCM0123-025GEA 43.78 CAD
TCM0123-025G TCM0123-500G TCM0123-100G
Hydroquinone monomethyl ether ≥99.0%
Hydroquinone monomethyl ether

CAS Number: 150-76-5
MDL Number: MFCD00002332
Molecular Formula: C7H8O2
Molecular Weight: 124.14
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 243
Melting point (°C): 57
Flash Point (°C): 128


Product Categories: Materials Chemistry, Polymerization Inhibitors, Building Blocks for Liquid Crystals, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: Hydroquinone Monomethyl Ether, 4-Hydroxyanisole, MEHQ

Formula: C₇H₈O₂
Boiling Pt: 243 °C
Melting Pt: 57 °C
Density: 1.55 g/cm³ (20 °C)
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00002332
CAS Number: 150-76-5
EINECS: 205-769-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=caa001b37accac1817252cecb826f0275cc1e125&vid=544ecda4328ea1407dc09a47267988f8c3428d73&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR