4-Nitro-o-anisidine ≥99.0%

Supplier: TCI America
Warning

Synonyms: 2-Methoxy-4-nitroaniline, 2-Amino-5-nitroanisole

M0118-500G M0118-25G
TCM0118-500GEA 287.56 CAD
TCM0118-500G TCM0118-025G
4-Nitro-o-anisidine ≥99.0%
4-Nitro-o-anisidine

CAS Number: 97-52-9
MDL Number: MFCD00007363
Molecular Formula: C7H8N2O3
Molecular Weight: 168.15
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 140


Other Product Names / Synonyms: 1-Amino-2-methoxy-4-nitrobenzene, 4-Nitro-o-anisidine

Formula: C₇H₈N₂O₃
Boiling Pt: 364 °C (1013 hPa)
Melting Pt: 140 °C
Density: 1.2112 g/cm³ (156 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007363
CAS Number: 97-52-9
EINECS: 202-588-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fcf775a6cc96453acc5e96658cebd6b04e92ee2&vid=5fcf775a6cc96453acc5e96658cebd6b04e92ee2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR