4-Methoxybenzyl alcohol ≥98.0%
Supplier: TCI America
|
Danger
|
M0107-500G
M0107-25G
M0107-100G
TCM0107-500GEA
205.4
CAD
TCM0107-500G
TCM0107-025G
TCM0107-100G
4-Methoxybenzyl alcohol ≥98.0%
4-Methoxybenzyl alcohol
CAS Number: 105-13-5
MDL Number: MFCD00004653
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 259
Flash Point (°C): 148
Freezing point (°C): 24
Other Product Names / Synonyms: Anise Alcohol, Anisyl Alcohol
Formula:
C₈H₁₀O₂ Boiling Pt: 259 °C Melting Pt: 20…24 °C Density: 1.1 g/cm³ (20 °C) Flash Pt: 148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004653 CAS Number: 105-13-5 EINECS: 203-273-6 Merck Index: 13,00608 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...