4'-Methoxyacetophenone ≥99.0%

Supplier: TCI America
Warning

Synonyms: Acetanisole, 4-Methoxyacetophenone

M0105-25G M0105-500G
TCM0105-025GEA 46.59 CAD
TCM0105-025G TCM0105-500G
4'-Methoxyacetophenone ≥99.0%
4'-Methoxyacetophenone

CAS Number: 100-06-1
MDL Number: MFCD00008745
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 260
Melting point (°C): 38
Flash Point (°C): 138
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Acetoanisole, 4-Acetylanisole

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 260 °C
Melting Pt: 38 °C
Density: 1.14 g/cm³ (20 °C)
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00008745
CAS Number: 100-06-1
EINECS: 202-815-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff019f382ac162cb9e8e4ddf4512dd879d60551d&vid=ff019f382ac162cb9e8e4ddf4512dd879d60551d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR