3'-Methoxyacetophenone ≥97.0%
Supplier: TCI America
Synonyms:
3′-Methoxyacetophenone
M0104-25ML
M0104-500ML
TCM0104-025MLEA
67.94
CAD
TCM0104-025ML
TCM0104-500ML
3'-Methoxyacetophenone ≥97.0%
3'-Methoxyacetophenone
CAS Number: 586-37-8
MDL Number: MFCD00008736
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 240
Flash Point (°C): 110
Specific Gravity (20/20): 1.10
Other Product Names / Synonyms: 3-Acetoanisole, 3-Acetylanisole
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Boiling Pt: 240 °C Melting Pt: –8 °C Density: 1.10 (20 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008736 CAS Number: 586-37-8 EINECS: 209-573-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...