Methyl red 0.1% in ethanol 95%

Supplier: TCI America
Danger

Synonyms: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid

M0067-100ML M0067-500ML
TCM0067-100MLEA 80.2 CAD
TCM0067-100ML TCM0067-500ML
Methyl red 0.1% in ethanol 95%
Methyl red

[for Titration]
CAS Number: 493-52-7
MDL Number: MFCD00002425
Molecular Weight: 269.30
Form: Clear Liquid
Color: Red
Flash Point (°C): 17

Formula: C₁₅H₁₅N₃O₂
MW: 269.3 g/mol
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00002425
CAS Number: 493-52-7
EINECS: 207-776-1
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49d5aa6373e380eeae8c12aa2c3e0d6c6b12040b&vid=bc6cb8772f7a3e35cc14fcb23f6c73864dfa1ae6&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR