Sodium mercaptoacetate (sodium thioglycolate) ≥93.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Thioglycolic Acid, Thioglycolic Acid Sodium Salt, Mercaptoacetic acid, sodium salt

M0053-25G M0053-500G M0053-100G
TCM0053-025GEA 43.39 CAD
TCM0053-025G TCM0053-500G TCM0053-100G
Sodium mercaptoacetate (sodium thioglycolate) ≥93.0% (by titrimetric analysis)
Sodium mercaptoacetate (Sodium thioglycolate)

CAS Number: 367-51-1
MDL Number: MFCD00043386
Molecular Formula: C2H4O2S
Molecular Weight: 114.09
Purity/Analysis Method: >93.0% (T)
Form: Crystal
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Mercaptoacetic Acid Sodium Salt, Sodium Mercaptoacetate, Thioglycolic Acid Sodium Salt

Formula: HSCH₂COONa
MW: 114.1 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00043386
CAS Number: 367-51-1
EINECS: 206-696-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5c76d6349070ac9d10e331a01a8e99d3e7fe8c5&vid=b5c76d6349070ac9d10e331a01a8e99d3e7fe8c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR