D-(-)-Mannitol ≥99.0%

Supplier: TCI America

Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol

M0044-25G M0044-500G
TCM0044-025GEA 41.79 CAD
TCM0044-025G TCM0044-500G
D-(-)-Mannitol ≥99.0%
D(-)-Mannitol

CAS Number: 69-65-8
MDL Number: MFCD00064287
Molecular Formula: C6H14O6
Molecular Weight: 182.17
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 168

Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Boiling Pt: 290…295 °C (4 hPa)
Melting Pt: 168 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064287
CAS Number: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cdb4155d1e93c00d660c6ae9cb6d94c497d1d44b&vid=cdb4155d1e93c00d660c6ae9cb6d94c497d1d44b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR