Manganese(III) acetylacetonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Manganese (III) tris(2,4-pentanedionato-O,O'), Tris(2,4-pentanedionato-O,O')manganese (III), Manganese (III) 2,4-pentanedionate

M0043-500G M0043-25G M0043-100G
TCM0043-500GEA 698.36 CAD
TCM0043-500G TCM0043-025G TCM0043-100G
Manganese(III) acetylacetonate ≥98.0% (by titrimetric analysis)
Manganese (III) acetylacetonate

CAS Number: 14284-89-0
MDL Number: MFCD00000023
Molecular Formula: C15H21MnO6
Molecular Weight: 352.27
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 161


Product Categories: Materials Chemistry, Magnetic Metal Complexes (Magnetic Materials)
Other Product Names / Synonyms: Acetylacetone Manganese(III) Salt, Manganese(III) Acetylacetonate

Formula: C₁₅H₂₁MnO₆
MW: 352.27 g/mol
Melting Pt: 161 °C
MDL Number: MFCD00000023
CAS Number: 14284-89-0
EINECS: 238-188-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f789d7a32393827116e889f1d451fd9034580747&vid=f789d7a32393827116e889f1d451fd9034580747&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR