(±)-Methyl mandelate ≥98.0%
Supplier: TCI America
M0040-500G
M0040-25G
TCM0040-500GEA
562.48
CAD
TCM0040-500G
TCM0040-025G
(±)-Methyl mandelate ≥98.0%
(±)-Methyl mandelate
CAS Number: 4358-87-6
MDL Number: MFCD00004493
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 250
Melting point (°C): 56
Other Product Names / Synonyms: DL-Mandelic Acid Methyl Ester
Formula:
C₉H₁₀O₃ Boiling Pt: 250 °C Melting Pt: 56 °C Density: 1.1756 g/cm³ (20 °C) |
MDL Number:
MFCD00004493 CAS Number: 4358-87-6 EINECS: 224-434-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...