Disodium malonate ≥99.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Sodium malonate dibasic, Malonic acid disodium salt
M0031-25G
M0031-500G
TCM0031-25GEA
71.04
CAD
TCM0031-25G
TCM0031-500G
Disodium malonate ≥99.0% (by titrimetric analysis)
Disodium malonate
CAS Number: 141-95-7
MDL Number: MFCD00002708
Molecular Formula: C3H4O4
Molecular Weight: 148.02
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Other Product Names / Synonyms: Malonic Acid Disodium Salt
Formula:
C₃H₂Na₂O₄ MW: 148.03 g/mol |
MDL Number:
MFCD00002708 CAS Number: 141-95-7 EINECS: 205-514-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...