Dimethyl malonate ≥98.0%

Supplier: TCI America

M0030-25G M0030-500G
TCM0030-025GEA 45.82 CAD
TCM0030-025G TCM0030-500G
Dimethyl malonate ≥98.0%
Dimethyl malonate

CAS Number: 108-59-8
MDL Number: MFCD00008460
Molecular Formula: C5H8O4
Molecular Weight: 132.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 184
Melting point (°C): -62
Flash Point (°C): 91
Specific Gravity (20/20): 1.16


Other Product Names / Synonyms: Malonic Acid Dimethyl Ester, Methyl Malonate

Formula: C₅H₈O₄
Boiling Pt: 184 °C
Melting Pt: –62 °C
Density: 1.16 (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00008460
CAS Number: 108-59-8
EINECS: 203-597-8
Merck Index: 12,06169
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbbd3282b921f0374e4e84afa0734471194233b1&vid=dbbd3282b921f0374e4e84afa0734471194233b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR