Diethyl malonate ≥99.0%

Supplier: TCI America

Synonyms: 1,3-Propanedioic acid diethyl ester, Malonic acid diethyl ester

M0029-500ML M0029-25ML
TCM0029-500MLEA 78.89 CAD
TCM0029-500ML TCM0029-025ML
Diethyl malonate ≥99.0%
Diethyl malonate

CAS Number: 105-53-3
MDL Number: MFCD00009195
Molecular Formula: C7H12O4
Molecular Weight: 160.17
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 199
Melting point (°C): -50
Flash Point (°C): 93
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: Malonic Acid Diethyl Ester, Ethyl Malonate

Formula: C₇H₁₂O₄
Boiling Pt: 199 °C
Melting Pt: –50 °C
Density: 1.06 (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00009195
CAS Number: 105-53-3
EINECS: 203-305-9
Merck Index: 13,03856

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR