Malonic acid ≥99.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
M0028-25G
M0028-500G
TCM0028-25GEA
43.17
CAD
TCM0028-25G
TCM0028-500G
Malonic acid ≥99.0% (by titrimetric analysis)
Malonic acid
CAS Number: 141-82-2
MDL Number: MFCD00002707
Molecular Formula: C3H4O4
Molecular Weight: 104.06
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 134
Flash Point (°C): 157
Other Product Names / Synonyms: Propanedioic Acid
Formula:
C₃H₄O₄ Boiling Pt: 387 °C (1013 hPa) Melting Pt: 134 °C Density: 1.669 g/cm³ (20 °C) Flash Pt: 157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002707 CAS Number: 141-82-2 EINECS: 205-503-0 Merck Index: 13,05732 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...