L(-)-Malic acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: L(-)-Malic acid, (S)-(-)-Hydroxy-1,4-butanedioic acid, (S)-(-)-Hydroxysuccinic acid, (S)-2-hydroxysuccinic acid, L(-)-Hydroxysuccinic acid, L(-)-Hydroxy-1,4-butanedioic acid, L-(–)-Malic Acid

M0022-25G M0022-500G M0022-100G
TCM0022-025GEA 58.7 CAD
TCM0022-025G TCM0022-500G TCM0022-100G
L(-)-Malic acid ≥98.0%
L(-)-Malic acid

CAS Number: 97-67-6
MDL Number: MFCD00064213
Molecular Formula: C4H6O5
Molecular Weight: 134.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 103
Flash Point (°C): 220
Specific rotation [a]20/D: -6.5 deg (C=10, Acetone)


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Chiral Building Blocks
Other Product Names / Synonyms: L-(-)-Apple Acid

Formula: C₄H₆O₅
MW: 134.09 g/mol
Boiling Pt: 322
Melting Pt: 103 °C
Density: 1.595 g/cm³ (20 °C)
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00064213
CAS Number: 97-67-6
EINECS: 202-601-5
Merck Index: 13,05730
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73bbec2cd4c0257940f17cd2626017e76670dd39&vid=73bbec2cd4c0257940f17cd2626017e76670dd39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR