Diethyl maleate ≥90.0%

Supplier: TCI America
Warning

Synonyms: Maleic acid diethyl ester, Diethyl (Z)-2-butenedioate

M0010-25ML M0010-500ML
TCM0010-025MLEA 30.02 CAD
TCM0010-025ML TCM0010-500ML
Diethyl maleate ≥90.0%
Diethyl maleate

CAS Number: 141-05-9
MDL Number: MFCD00009191
Molecular Formula: C8H12O4
Molecular Weight: 172.18
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 225
Melting point (°C): -10
Flash Point (°C): 107
Specific Gravity (20/20): 1.07


Product Categories: Materials Chemistry, Resin Additives, Copolymerization Monomers
Other Product Names / Synonyms: Maleic Acid Diethyl Ester, Ethyl Maleate

Formula: C₈H₁₂O₄
Boiling Pt: 225 °C
Melting Pt: –10 °C
Density: 1.07 (20 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00009191
CAS Number: 141-05-9
EINECS: 205-451-9
Merck Index: 13,03150
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a924600a8f4016093dd37e86f61e14f908306196&vid=a924600a8f4016093dd37e86f61e14f908306196&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR