Lithium bromide ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Warning

L0210-25G L0210-500G L0210-100G
TCL0210-25GEA 200.33 CAD
TCL0210-25G TCL0210-500G TCL0210-100G
Lithium bromide ≥99.0% (by titrimetric analysis)
Lithium bromide

CAS Number: 7550-35-8
MDL Number: MFCD00011077
Molecular Formula: BrLi
Molecular Weight: 86.84
Purity/Analysis Method: >99.0% (T)
Form: Powder
Melting point (°C): 549

Formula: LiBr
Boiling Pt: 1265 °C (1013 hPa)
Melting Pt: 549 °C
Density: 3.46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011077
CAS Number: 7550-35-8
EINECS: 231-439-8
Merck Index: 13,05548
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=602b1c0bfcd8eea4419c70ecec2f1ce1c83c81a7&vid=602b1c0bfcd8eea4419c70ecec2f1ce1c83c81a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR