Ethyl levulinate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 4-oxopentanoate, 4-Oxovaleric acid ethyl ester, Ethyl 4-oxovalerate, 4-Ketovaleric acid ethyl ester, 4-Oxopentanoic acid ethyl ester, Levulinic acid ethyl ester

L0120-25ML L0120-500ML L0120-100ML
TCL0120-025MLEA 35.34 CAD
TCL0120-025ML TCL0120-500ML TCL0120-100ML
Ethyl levulinate ≥98.0%
Ethyl levulinate

CAS Number: 539-88-8
MDL Number: MFCD00009209
Molecular Formula: C7H12O3
Molecular Weight: 144.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 100
Specific Gravity (20/20): 1.01


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: 3-Acetylpropionic Acid Ethyl Ester, 4-Ketovaleric Acid Ethyl Ester, Levulinic Acid Ethyl Ester, 4-Oxovaleric Acid Ethyl Ester

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 205 °C
Density: 1.01 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00009209
CAS Number: 539-88-8
EINECS: 208-728-2
Merck Index: 13,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55dcd17cabb256540674f164a8929df989dc4060&vid=55dcd17cabb256540674f164a8929df989dc4060&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR