(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: L-Lactide, (3S,6S)-3,6-Dimethyl-1,4-dioxane-2,5-dione

L0115-250G L0115-25G
TCL0115-250GEA 881.64 CAD
TCL0115-250G TCL0115-025G
(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione ≥98.0% (by titrimetric analysis)
(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione

CAS Number: 4511-42-6
MDL Number: MFCD00070594
Molecular Formula: C6H8O4
Molecular Weight: 144.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 97
Flash Point (°C): 200
Specific rotation [a]20/D: -290 deg (C=1, Toluene)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Materials Chemistry, Monomers, Bioscience, Monomers for Synthetic Polymers (for DDS)
Other Product Names / Synonyms: L-(-)-Dilactide, (3S,6S)-(-)-3,6-Dimethyl-1,4-dioxane-2,5-dione

Formula: C₆H₈O₄
MW: 144.13 g/mol
Melting Pt: 97 °C
Flash Pt: 200 °C
MDL Number: MFCD00070594
CAS Number: 4511-42-6
EINECS: 224-832-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00f184c8c9c5f2edfc06360e85488cd122881d68&vid=00f184c8c9c5f2edfc06360e85488cd122881d68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR