Dodecanenitrile ≥98.0%
Supplier: TCI America
L0110-500ML
L0110-25ML
TCL0110-500MLEA
477.16
CAD
TCL0110-500ML
TCL0110-025ML
Dodecanenitrile ≥98.0%
Dodecanenitrile
CAS Number: 2437-25-4
MDL Number: MFCD00001946
Molecular Formula: C12H23N
Molecular Weight: 181.32
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 276
Flash Point (°C): 114
Specific Gravity (20/20): 0.83
Other Product Names / Synonyms: Dodecanenitrile, Undecyl Cyanide
Formula:
C₁₂H₂₃N Boiling Pt: 276 °C Density: 0.83 (20 °C) Flash Pt: 114 °C |
MDL Number:
MFCD00001946 CAS Number: 2437-25-4 EINECS: 219-440-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...