Butyl laurate ≥99.0%

Supplier: TCI America

Synonyms: Butyl dodecanoate, Dodecylic acid butyl ester, Dodecanoic acid butyl ester, Lauric acid butyl ester, n-Butyl dodecanoate

L0083-500ML L0083-25ML
TCL0083-500MLEA 271.76 CAD
TCL0083-500ML TCL0083-025ML
Butyl laurate ≥99.0%
Butyl laurate

CAS Number: 106-18-3
MDL Number: MFCD00042870
Molecular Formula: C16H32O2
Molecular Weight: 256.43
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 199
Flash Point (°C): 166
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: Lauric Acid Butyl Ester

Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
Boiling Pt: 199 °C
Density: 0.86 (20 °C)
Flash Pt: 166 °C
MDL Number: MFCD00042870
CAS Number: 106-18-3
EINECS: 203-370-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a96a1f09c0c78a2bae1137aafd2daabfd61a187a&vid=a96a1f09c0c78a2bae1137aafd2daabfd61a187a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR