Methyl linoleate ≥95.0%
Supplier: TCI America
L0078-25ML
L0078-500ML
TCL0078-025MLEA
181.86
CAD
TCL0078-025ML
TCL0078-500ML
Methyl linoleate ≥95.0%
Methyl linoleate
CAS Number: 112-63-0
MDL Number: MFCD00009534
Molecular Formula: C19H34O2
Molecular Weight: 294.48
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 346
Melting point (°C): -35
Flash Point (°C): 200
Specific Gravity (20/20): 0.89
Product Categories: Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Linoleic Acid Methyl Ester
Formula:
C₁₉H₃₄O₂ Boiling Pt: 346 °C Melting Pt: –35 °C Density: 0.89 (20 °C) Flash Pt: 200 °C |
MDL Number:
MFCD00009534 CAS Number: 112-63-0 EINECS: 203-993-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...