L(+)-Lysine monohydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Lysine Hydrochloride, (S)-(+)-2,6-Diaminohexanoic acid monohydrochloride, L-Lysine monohydrochloride, H-Lys-OH·HCl, L-Lysine monohydrocholoride, L-Lysine Hydrochloride, (S)-(+)-Lysine monohydrochloride, (S)-(+)-2,6-Diaminocaproic acid monohydrochloride, H-Lys-OH.HCl

L0071-25G L0071-500G
TCL0071-025GEA 62.69 CAD
TCL0071-025G TCL0071-500G
L(+)-Lysine monohydrochloride ≥98.0% (by titrimetric analysis)
L(+)-Lysine monohydrochloride

CAS Number: 657-27-2
MDL Number: MFCD00064564
Molecular Formula: C6H14N2O2
Molecular Weight: 182.65
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 20 deg (C=8, HCl(1+1))


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-Lys-OH.HCl

Formula: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
MW: 182.65 g/mol
Melting Pt: 263…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00064564
CAS Number: 657-27-2
EINECS: 211-519-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f4385362f12dc8d5d151b69a4bc4a9cdadaaa53&vid=6f4385362f12dc8d5d151b69a4bc4a9cdadaaa53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR