DL-Lysine monohydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
L0070-100G
L0070-25G
TCL0070-100GEA
120.08
CAD
TCL0070-100G
TCL0070-025G
DL-Lysine monohydrochloride ≥98.0% (by titrimetric analysis)
DL-Lysine monohydrochloride
CAS Number: 70-53-1
MDL Number: MFCD00064563
Molecular Formula: C6H14N2O2
Molecular Weight: 182.65
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-DL-Lys-OH.HCl
Formula:
C₆H₁₅ClN₂O₂ Melting Pt: 267 °C |
MDL Number:
MFCD00064563 CAS Number: 70-53-1 EINECS: 200-739-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...