(±)-Linalool ≥96.0%

Supplier: TCI America
Warning

L0048-25ML L0048-500ML
TCL0048-025MLEA 63.33 CAD
TCL0048-025ML TCL0048-500ML
(±)-Linalool ≥96.0%
(±)-Linalool

CAS Number: 78-70-6
MDL Number: MFCD00008906
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 200
Flash Point (°C): 71
Specific Gravity (20/20): 0.86

Formula: C₁₀H₁₈O
Boiling Pt: 200 °C
Density: 0.86 (20 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00008906
CAS Number: 78-70-6
EINECS: 201-134-4
Merck Index: 13,05517
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5733b7430eaceba1185702c1393a094e38d967ab&vid=5733b7430eaceba1185702c1393a094e38d967ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR