Methyl levulinate ≥99.0%

Supplier: TCI America

L0044-25G L0044-500G L0044-100G
TCL0044-025GEA 60.04 CAD
TCL0044-025G TCL0044-500G TCL0044-100G
Methyl levulinate ≥99.0%
Methyl levulinate

CAS Number: 624-45-3
MDL Number: MFCD00017499
Molecular Formula: C6H10O3
Molecular Weight: 130.14
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 196
Flash Point (°C): 72
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: Levulinic Acid Methyl Ester, 4-Oxovaleric Acid Methyl Ester, Methyl 4-Oxovalerate, Methyl 4-Oxopentanoate

Formula: C₆H₁₀O₃
Boiling Pt: 196 °C
Melting Pt: –24 °C
Density: 1.05 (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00017499
CAS Number: 624-45-3
EINECS: 210-846-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4767e67abcd22e45b6e2a3c2db040d5f97065b7&vid=b4767e67abcd22e45b6e2a3c2db040d5f97065b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR