D-(-)-Leucine ≥98.0%

Supplier: TCI America

Synonyms: H-D-Leu-OH, (R)-(-)-Leucine

L0027-5G L0027-25G
TCL0027-005GEA 71.1 CAD
TCL0027-005G TCL0027-025G
D-(-)-Leucine ≥98.0%
D(-)-Leucine

CAS Number: 328-38-1
MDL Number: MFCD00063088
Molecular Formula: C6H13NO2
Molecular Weight: 131.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -15 deg (C=4, 6mol/L HCl)


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-D-Leu-OH

Formula: C₆H₁₃NO₂
Boiling Pt: 226 °C (1013 hPa)
MDL Number: MFCD00063088
CAS Number: 328-38-1
EINECS: 206-327-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ead5baabae411434bcb86fb41a44a18d70592e35&vid=ead5baabae411434bcb86fb41a44a18d70592e35&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR