4-Methylquinoline ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Lepidine

L0024-25ML L0024-500ML L0024-100ML
TCL0024-025MLEA 107.72 CAD
TCL0024-025ML TCL0024-500ML TCL0024-100ML
4-Methylquinoline ≥97.0% (by GC, titration analysis)
4-Methylquinoline

CAS Number: 491-35-0
MDL Number: MFCD00006784
Molecular Formula: C10H9N
Molecular Weight: 143.19
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 265
Melting point (°C): 10
Flash Point (°C): 107
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: 4-Methylquinoline

Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 265 °C
Melting Pt: 10 °C
Density: 1.08 (20 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00006784
CAS Number: 491-35-0
EINECS: 207-734-2
Merck Index: 13,05460
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66ce4acf71b78e87a46344c7899059351d05429c&vid=336310ce87fb4db6f286cdab5eaef6cf77769f1c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR