Lecithin Soy Bean

Supplier: TCI America

L0023-25G L0023-500G
TCL0023-25GEA 59.77 CAD
TCL0023-25G TCL0023-500G
Lecithin Soy Bean
Lecithin

CAS Number: 8002-43-5
MDL Number: MFCD00147406
Form: Clear Liquid
Color: Yellow


Product Categories: Bioscience, Lipids

MDL Number: MFCD00147406
CAS Number: 8002-43-5
EINECS: 310-129-7
Merck Index: 12,05452

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR